Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSSC4
Entrez Gene ID
705185
Gene Name
tumor suppressing subtransferable candidate 4
Gene Aliases
EGK_05871
Chromosome Location
Chr.14: 2462088 - 2467591 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193980.2 NP_001180909.2 - 339 144
XM_001093442.4 XP_001093442.3 - 2544 144
XM_028832628.1 XP_028688461.1 - 1324 144
GenBank mRNA JU332394.1 - - 160 144
JU472934.1 - - 164 144

View Full Product Details