Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TXNDC9
Entrez Gene ID
709303
Gene Name
thioredoxin domain containing 9
Gene Aliases
EGK_05607
Chromosome Location
Chr.13: 3141315 - 3156630 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194267.2 NP_001181196.1 - 221 121
XM_001104266.4 XP_001104266.1 - 445 121
XM_015112908.2 XP_014968394.1 - 297 121
GenBank mRNA JU332441.1 - - 362 121
JU472973.1 - - 351 121
JV045208.1 - - 323 121

View Full Product Details