Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FOXO4
Entrez Gene ID
698055
Gene Name
forkhead box O4
Gene Aliases
EGK_20618
Chromosome Location
Chr.X: 68083007 - 68090731 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015127651.2 XP_014983137.1 - 2213 142
GenBank mRNA JU334572.1 - - 1893 142
JU475655.1 - - 1899 142
JV047105.1 - - 1899 142

View Full Product Details