Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACTL7B
Entrez Gene ID
710750
Gene Name
actin like 7B
Gene Aliases
EGK_07454
Chromosome Location
Chr.15: 34050914 - 34058950 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001108601.3 XP_001108601.1 - 1150 112

View Full Product Details