Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CLDN14
Entrez Gene ID
695087
Gene Name
claudin 14
Gene Aliases
-
Chromosome Location
Chr.3: 10114294 - 10134497 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193733.3 NP_001180662.2 - 644 78
XM_015132885.2 XP_014988371.2 - 1343 78
XM_028844799.1 XP_028700632.1 - 1444 78
XM_028844800.1 XP_028700633.1 - 1568 78
XM_028844801.1 XP_028700634.1 - 1327 78

View Full Product Details