Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SUGP1
Entrez Gene ID
719772
Gene Name
SURP and G-patch domain containing 1
Gene Aliases
SF4
Chromosome Location
Chr.19: 18872859 - 18919028 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261744.2 NP_001248673.1 - 211 124
XM_015123385.2 XP_014978871.1 - 547 124
GenBank mRNA JU321359.1 - - 209 124
JV044634.1 - - 212 124

View Full Product Details