Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SIGLEC6
Entrez Gene ID
719684
Gene Name
sialic acid binding Ig like lectin 6
Gene Aliases
-
Chromosome Location
Chr.19: 51295174 - 51311208 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015124709.2 XP_014980195.2 - 866 153

View Full Product Details