Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FOXI2
Entrez Gene ID
703280
Gene Name
forkhead box I2
Gene Aliases
-
Chromosome Location
Chr.9: 128180396 - 128182211 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001091576.4 XP_001091576.2 - 598 136

View Full Product Details