Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RPS4X
Entrez Gene ID
699397
Gene Name
ribosomal protein S4 X-linked
Gene Aliases
-
Chromosome Location
Chr.X: 69346879 - 69351493 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001265622.1 NP_001252551.1 - 79 150
GenBank mRNA JV667304.1 - - 79 150

View Full Product Details