Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ZIC2
- Entrez Gene ID
-
705399
- Gene Name
- Zic family member 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.17: 81696707 - 81701686 on Build Mmul_10
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
JV627950.1 |
- |
- |
976 |
148 |
JV627952.1 |
- |
- |
976 |
148 |
- Gene Symbol
- ZIC1
- Entrez Gene ID
-
715102
- Gene Name
- Zic family member 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.2: 49542251 - 49547537 on Build Mmul_10
View Full Product Details