Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRKRA
Entrez Gene ID
703063
Gene Name
protein activator of interferon induced protein kinase EIF2AK2
Gene Aliases
-
Chromosome Location
Chr.12: 65561754 - 65582527 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260847.1 NP_001247776.1 - 428 155
GenBank mRNA JU336864.1 - - 425 155
JU477604.1 - - 1848 155
JV048545.1 - - 428 155

View Full Product Details