Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MIEN1
Entrez Gene ID
697837
Gene Name
migration and invasion enhancer 1
Gene Aliases
C16H17orf37
Chromosome Location
Chr.16: 50855239 - 50862810 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266804.1 NP_001253733.1 - 234 138
GenBank mRNA JU477494.1 - - 234 138

View Full Product Details