Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBE2V1
Entrez Gene ID
100425552
Gene Name
ubiquitin conjugating enzyme E2 V1
Gene Aliases
-
Chromosome Location
Chr.10: 85308070 - 85343750 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015148758.2 XP_015004244.1 - 1099 142
XM_028827088.1 XP_028682921.1 - 916 142
XM_028827089.1 XP_028682922.1 - 1090 142
XM_028827091.1 XP_028682924.1 - 1119 142
GenBank mRNA JU328418.1 - - 805 142
JU473144.1 - - 890 142
JV045237.1 - - 887 142

View Full Product Details