Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC100427847
Entrez Gene ID
100427847
Gene Name
PRAME family member 17-like
Gene Aliases
-
Chromosome Location
Chr.1: 211614476 - 211617871 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015110098.2 XP_014965584.2 - 1361 132

Gene Symbol
LOC715953
Entrez Gene ID
715953
Gene Name
PRAME family member 17
Gene Aliases
PRAMEF10
Chromosome Location
Chr.1: 211591725 - 211597334 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015110192.2 XP_014965678.2 - 1421 132

View Full Product Details