Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC114673061
Entrez Gene ID
114673061
Gene Name
heparan sulfate glucosamine 3-O-sulfotransferase 3B1
Gene Aliases
-
Chromosome Location
Chr.16: 14056375 - 14105740 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028836030.1 XP_028691863.1 - 184 79

View Full Product Details