Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBE2F
Entrez Gene ID
695394
Gene Name
ubiquitin conjugating enzyme E2 F (putative)
Gene Aliases
-
Chromosome Location
Chr.12: 125367792 - 125443523 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015111244.2 XP_014966730.1 - 1268 149
XM_028830396.1 XP_028686229.1 - 1347 149
XM_028830397.1 XP_028686230.1 - 1583 149
XM_028830398.1 XP_028686231.1 - 1338 149
XM_028830399.1 XP_028686232.1 - 1324 149
XM_028830400.1 XP_028686233.1 - 1303 149
XM_028830401.1 XP_028686234.1 - 1261 149
GenBank mRNA JU333026.1 - - 1193 149
JV707339.1 - - 1222 149

View Full Product Details