Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAM210A
Entrez Gene ID
701669
Gene Name
family with sequence similarity 210 member A
Gene Aliases
C18H18orf19, C18orf19
Chromosome Location
Chr.18: 74295159 - 74358204 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015121547.2 XP_014977033.1 - 1853 152
XM_028837720.1 XP_028693553.1 - 1725 152
XM_028837721.1 XP_028693554.1 - 1979 152
XM_028837722.1 XP_028693555.1 - 1672 152
GenBank mRNA JU332222.1 - - 1645 152

View Full Product Details