Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYP2B6
Entrez Gene ID
678687
Gene Name
cytochrome P450, family 2, subfamily B, polypeptide 6
Gene Aliases
CYP2B30
Chromosome Location
Chr.19: 117373 - 139783 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040212.1 NP_001035302.1 - 1147 87
GenBank mRNA AY635461.1 - - 1147 87

Gene Symbol
LOC114675607
Entrez Gene ID
114675607
Gene Name
cytochrome P450 2B6-like
Gene Aliases
-
Chromosome Location
Chr.19: 6235 - 28968 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028842827.1 XP_028698660.1 - 1196 87

View Full Product Details