Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYP1A1
Entrez Gene ID
678694
Gene Name
cytochrome P450, family 1, subfamily A, polypeptide 1
Gene Aliases
-
Chromosome Location
Chr.7: 51749579 - 51755598 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040238.1 NP_001035328.1 - 1249 67
XM_015142702.2 XP_014998188.2 - 1389 67
XM_015142703.2 XP_014998189.2 - 1387 67
GenBank mRNA AY635458.1 - - 1249 67

View Full Product Details