Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
APLNR
Entrez Gene ID
706823
Gene Name
apelin receptor
Gene Aliases
AGTRL1, APJ, EGK_06232
Chromosome Location
Chr.14: 16320228 - 16321370 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047126.1 NP_001040591.1 - 635 116
GenBank mRNA JV565075.1 - - 795 116
JV608024.1 - - 908 116

View Full Product Details