Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CXCR6
Entrez Gene ID
713829
Gene Name
C-X-C motif chemokine receptor 6
Gene Aliases
Bonzo, CD186, STRL33
Chromosome Location
Chr.2: 101948255 - 101953190 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047141.1 NP_001040606.1 - 962 79
XM_015131078.2 XP_014986564.1 - 1108 79
XM_015131079.2 XP_014986565.1 - 1647 79
GenBank mRNA DQ902400.1 - - 761 79
DQ902401.1 - - 741 79
DQ902402.1 - - 759 79
DQ902403.1 - - 759 79
DQ902404.1 - - 634 79
DQ902406.1 - - 760 79
DQ902407.1 - - 761 79
DQ902408.1 - - 761 79
DQ902409.1 - - 760 79
DQ902410.1 - - 724 79
DQ902411.1 - - 759 79
DQ902412.1 - - 754 79
DQ902413.1 - - 747 79
DQ902414.1 - - 760 79
DQ902415.1 - - 754 79
DQ902416.1 - - 760 79
DQ902417.1 - - 761 79
DQ902418.1 - - 759 79
DQ902419.1 - - 760 79
DQ902420.1 - - 701 79
DQ902421.1 - - 761 79
DQ902423.1 - - 752 79
DQ902424.1 - - 761 79
DQ902425.1 - - 760 79
DQ902426.1 - - 752 79
DQ902428.1 - - 760 79
DQ902429.1 - - 754 79
DQ902430.1 - - 756 79
DQ902431.1 - - 753 79
DQ902432.1 - - 761 79
DQ902433.1 - - 761 79
DQ902434.1 - - 760 79
DQ902435.1 - - 760 79
DQ902436.1 - - 695 79
DQ902437.1 - - 758 79
DQ902438.1 - - 760 79
DQ902439.1 - - 760 79
DQ902440.1 - - 761 79
JU474040.1 - - 1007 79

View Full Product Details