Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KIR3DL0
Entrez Gene ID
696175
Gene Name
killer immunoglobulin-like receptor KIR3DL0
Gene Aliases
-
Chromosome Location
Chr.19: 54355344 - 54365976 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047119.2 NP_001040584.1 - 39 70
GenBank mRNA DQ157756.1 - - 40 70

View Full Product Details