Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HAS3
Entrez Gene ID
704121
Gene Name
hyaluronan synthase 3
Gene Aliases
-
Chromosome Location
Chr.20: 56337489 - 56368224 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110777.2 NP_001104247.2 - 1341 79
XM_028840467.1 XP_028696300.1 - 1227 79
XM_028840468.1 XP_028696301.1 - 1757 79
GenBank mRNA JU485307.1 - - 689 79
JV649669.1 - - 409 79

View Full Product Details