Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NLRP2
Entrez Gene ID
697844
Gene Name
NLR family pyrin domain containing 2
Gene Aliases
NALP2
Chromosome Location
Chr.19: 54699186 - 54725219 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114352.1 NP_001107824.1 - 274 108
GenBank mRNA EU344967.1 - - 274 108
JV655647.1 - - 330 108

View Full Product Details