Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DIO1
Entrez Gene ID
714739
Gene Name
iodothyronine deiodinase 1
Gene Aliases
-
Chromosome Location
Chr.1: 170508277 - 170525620 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122652.3 NP_001116124.1 - 360 80

View Full Product Details