Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDE9A
Entrez Gene ID
100301991
Gene Name
phosphodiesterase 9A
Gene Aliases
PDE9A2
Chromosome Location
Chr.3: 3888596 - 4009128 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161449.1 NP_001154921.1 - 1330 98
XM_028844729.1 XP_028700562.1 - 1685 98
XM_028844730.1 XP_028700563.1 - 1418 98
XM_028844731.1 XP_028700564.1 - 1007 98
GenBank mRNA FJ914626.1 - - 1330 98
JU321678.1 - - 1332 98
JU475918.1 - - 1323 98
JV584082.1 - - 592 98

View Full Product Details