Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDE9A
Entrez Gene ID
100301991
Gene Name
phosphodiesterase 9A
Gene Aliases
PDE9A2
Chromosome Location
Chr.3: 3888596 - 4009128 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161449.1 NP_001154921.1 - 314 57
GenBank mRNA FJ914626.1 - - 314 57
JU321678.1 - - 316 57
JU475918.1 - - 307 57

View Full Product Details