Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LCN8
Entrez Gene ID
709395
Gene Name
lipocalin 8
Gene Aliases
ESP20.5
Chromosome Location
Chr.15: 1469663 - 1473657 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171840.1 NP_001165311.1 - 505 183
GenBank mRNA AF218048.1 - - 505 183

View Full Product Details