Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MIR181C
Entrez Gene ID
100315138
Gene Name
microRNA mir-181c
Gene Aliases
mir-181c, mml-mir-181c
Chromosome Location
Chr.19: 13475956 - 13476065 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_032033.1 - 59 58

View Full Product Details