Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HIST1H2BH
Entrez Gene ID
698482
Gene Name
histone cluster 1, H2bh
Gene Aliases
-
Chromosome Location
Chr.4: 143735108 - 143743725 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193825.1 NP_001180754.1 - 527 78

View Full Product Details