Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GXYLT1
Entrez Gene ID
698940
Gene Name
glucoside xylosyltransferase 1
Gene Aliases
GLT8D3
Chromosome Location
Chr.11: 41474747 - 41536884 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193857.2 NP_001180786.2 - 725 107
XM_015151455.2 XP_015006941.2 - 505 107
GenBank mRNA JV648758.1 - - 306 107

View Full Product Details