Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TENM3
Entrez Gene ID
700867
Gene Name
teneurin transmembrane protein 3
Gene Aliases
ODZ1, ODZ3
Chromosome Location
Chr.5: 178767099 - 180127370 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193929.2 NP_001180858.2 - 3478 80
XM_028848051.1 XP_028703884.1 - 3828 80
XM_028848052.1 XP_028703885.1 - 3550 80
XM_028848053.1 XP_028703886.1 - 3825 80
XM_028848054.1 XP_028703887.1 - 3828 80
XM_028848055.1 XP_028703888.1 - 3807 80
XM_028848056.1 XP_028703889.1 - 3828 80
XM_028848057.1 XP_028703890.1 - 3828 80
XM_028848058.1 XP_028703891.1 - 3780 80
XM_028848059.1 XP_028703892.1 - 3780 80
XM_028848060.1 XP_028703893.1 - 3264 80
XM_028848061.1 XP_028703894.1 - 2954 80

View Full Product Details