Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HIST1H2BJ
Entrez Gene ID
701337
Gene Name
histone cluster 1, H2bj
Gene Aliases
-
Chromosome Location
Chr.4: 143081678 - 143088084 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193949.1 NP_001180878.1 - 388 71

View Full Product Details