Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NUDT7
Entrez Gene ID
715443
Gene Name
nudix hydrolase 7
Gene Aliases
EGK_13026
Chromosome Location
Chr.20: 64773412 - 64792788 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194315.1 NP_001181244.1 - 99 69
XM_015126656.2 XP_014982142.1 - 123 69
XM_015126657.2 XP_014982143.1 - 123 69
XM_028840581.1 XP_028696414.1 - 123 69
GenBank mRNA JU323580.1 - - 97 69

View Full Product Details