Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
INO80E
Entrez Gene ID
708115
Gene Name
INO80 complex subunit E
Gene Aliases
-
Chromosome Location
Chr.20: 27358570 - 27368014 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194364.2 NP_001181293.1 - 163 86
GenBank mRNA JU471949.1 - - 186 86
JV712552.1 - - 163 86

View Full Product Details