Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RHOG
Entrez Gene ID
718113
Gene Name
ras homolog family member G
Gene Aliases
-
Chromosome Location
Chr.14: 64615716 - 64630149 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194601.2 NP_001181530.1 - 277 100

View Full Product Details