Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RFXANK
Entrez Gene ID
719706
Gene Name
regulatory factor X associated ankyrin containing protein
Gene Aliases
EGK_10354
Chromosome Location
Chr.19: 18781979 - 18793856 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194670.2 NP_001181599.1 - 977 89
XM_015123382.2 XP_014978868.1 - 1038 89
XM_015123383.2 XP_014978869.1 - 1032 89
XM_015123384.2 XP_014978870.1 - 1039 89
XM_028838541.1 XP_028694374.1 - 1046 89
XM_028838542.1 XP_028694375.1 - 871 89
XM_028838543.1 XP_028694376.1 - 1040 89
XM_028838544.1 XP_028694377.1 - 1042 89
GenBank mRNA JU336876.1 - - 708 89
JU477613.1 - - 720 89
JV048549.1 - - 718 89
JV566930.1 - - 718 89
JV664271.1 - - 740 89

View Full Product Details