Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- DDIT3
- Entrez Gene ID
-
714901
- Gene Name
- DNA damage inducible transcript 3
- Gene Aliases
-
EGK_03839
- Chromosome Location
-
Chr.11: 57091125 - 57095818 on Build Mmul_10
- Gene Symbol
- MARS1
- Entrez Gene ID
-
714744
- Gene Name
- methionyl-tRNA synthetase 1
- Gene Aliases
-
MARS
- Chromosome Location
-
Chr.11: 57064070 - 57091273 on Build Mmul_10
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
JU322390.1 |
- |
- |
3323 |
92 |
JV044407.1 |
- |
- |
3312 |
92 |
View Full Product Details