Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAM219B
Entrez Gene ID
722132
Gene Name
family with sequence similarity 219 member B
Gene Aliases
C15orf17
Chromosome Location
Chr.7: 51943625 - 51951753 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194738.2 NP_001181667.2 - 457 59
XM_015142708.2 XP_014998194.2 - 887 59
XM_015142710.2 XP_014998196.2 - 887 59
XM_015142711.2 XP_014998197.2 - 887 59
GenBank mRNA JU322524.1 - - 496 59
JV638133.1 - - 418 59

View Full Product Details