Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SURF2
Entrez Gene ID
100430364
Gene Name
surfeit 2
Gene Aliases
-
Chromosome Location
Chr.15: 4895650 - 4900982 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194794.3 NP_001181723.2 - 382 68
XM_015116406.2 XP_014971892.1 - 615 68
GenBank mRNA JU323404.1 - - 392 68
JU473240.1 - - 376 68
JV529498.1 - - 388 68
JV565037.1 - - 383 68
JV704655.1 - - 385 68

View Full Product Details