Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CFC1
Entrez Gene ID
702829
Gene Name
cripto, FRL-1, cryptic family 1
Gene Aliases
-
Chromosome Location
Chr.12: 394028 - 427396 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190851.2 NP_001177780.2 - 242 88

View Full Product Details