Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYCE3
Entrez Gene ID
720691
Gene Name
synaptonemal complex central element protein 3
Gene Aliases
C10H22orf41, C22orf41
Chromosome Location
Chr.10: 924875 - 938512 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193353.1 NP_001180282.1 - 110 102
XM_015150640.2 XP_015006126.2 - 242 102
XM_015150641.2 XP_015006127.1 - 217 102
XM_015150642.2 XP_015006128.1 - 222 102
XM_015150643.2 XP_015006129.1 - 129 102
XM_028827466.1 XP_028683299.1 - 143 102
GenBank mRNA JU321451.1 - - 154 102
JV048995.1 - - 153 102

View Full Product Details