Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HTR1F
Entrez Gene ID
693418
Gene Name
5-hydroxytryptamine receptor 1F
Gene Aliases
-
Chromosome Location
Chr.2: 184065324 - 184263405 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193683.2 NP_001180612.2 - 1044 100
XM_015128504.2 XP_014983990.2 - 1547 100
XM_028843054.1 XP_028698887.1 - 1846 100
XM_028843055.1 XP_028698888.1 - 1653 100
XM_028843056.1 XP_028698889.1 - 1621 100
XM_028843057.1 XP_028698890.1 - 1574 100
XM_028843058.1 XP_028698891.1 - 1576 100

View Full Product Details