Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLEKHF1
Entrez Gene ID
700343
Gene Name
pleckstrin homology and FYVE domain containing 1
Gene Aliases
EGK_10407
Chromosome Location
Chr.19: 29108359 - 29118562 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193737.2 NP_001180666.1 - 85 76
GenBank mRNA JU493312.1 - - 66 76

View Full Product Details