Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IQCK
Entrez Gene ID
695417
Gene Name
IQ motif containing K
Gene Aliases
-
Chromosome Location
Chr.20: 18549286 - 18686722 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193745.3 NP_001180674.1 - 426 92
XM_015125656.2 XP_014981142.1 - 443 92
XM_028840372.1 XP_028696205.1 - 307 92
XM_028840373.1 XP_028696206.1 - 426 92
GenBank mRNA JU325458.1 - - 253 92
JV051454.1 - - 278 92

View Full Product Details