Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NDUFA5
Entrez Gene ID
100430178
Gene Name
NADH:ubiquinone oxidoreductase subunit A5
Gene Aliases
-
Chromosome Location
Chr.3: 149962653 - 149980432 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198685.2 NP_001185614.1 - 29 59
GenBank mRNA CB229416.1 - - 32 59
JV044471.1 - - 32 59
JV044472.1 - - 32 59

View Full Product Details