Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TPSG1
Entrez Gene ID
100425751
Gene Name
tryptase gamma 1
Gene Aliases
-
Chromosome Location
Chr.20: 1219704 - 1223261 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204311.2 NP_001191240.2 - 522 87
XM_028840325.1 XP_028696158.1 - 543 87

View Full Product Details