Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAJC25
Entrez Gene ID
708100
Gene Name
DnaJ (Hsp40) homolog, subfamily C , member 25
Gene Aliases
-
Chromosome Location
Chr.15: 31169951 - 31190908 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204971.1 NP_001191900.1 - 1011 94
GenBank mRNA CO645394.1 - - 472 94

View Full Product Details