Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OTX2
Entrez Gene ID
698501
Gene Name
orthodenticle homeobox 2
Gene Aliases
EGK_18218
Chromosome Location
Chr.7: 118901435 - 118912050 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177645.1 NP_001171116.1 - 100 83
XM_015143755.2 XP_014999241.1 - 395 83
XM_028850243.1 XP_028706076.1 - 292 83

View Full Product Details