Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NECAP1
Entrez Gene ID
716035
Gene Name
NECAP endocytosis associated 1
Gene Aliases
EGK_03324
Chromosome Location
Chr.11: 11419365 - 11435481 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194593.2 NP_001181522.1 - 392 94
GenBank mRNA JU333891.1 - - 456 94
JU475076.1 - - 400 94
JV044876.1 - - 409 94
JV669285.1 - - 467 94

View Full Product Details